(4-Nitrophenyl)acetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Nitrophenylacetic acid

N0227-25G N0227-500G
TCN0227-025GEA 35 USD
TCN0227-025G TCN0227-500G
(4-Nitrophenyl)acetic acid ≥98.0% (by HPLC, titration analysis)
(4-Nitrophenyl)acetic acid

CAS Number: 104-03-0
MDL Number: MFCD00007383
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 153

Formula: (C₆H₄(NO₂))CH₂COOH
MW: 181.15 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00007383
CAS Number: 104-03-0
EINECS: 203-168-5
Merck Index: 12,06719
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=489095b970a865a57cee5c8b8cfd097bc2e62d39&vid=489095b970a865a57cee5c8b8cfd097bc2e62d39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR