(2-Nitrophenoxy)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(2-Nitrophenoxy)acetic acid

N0225-5G N0225-25G
TCN0225-5GEA 73.2 USD
TCN0225-5G TCN0225-025G
(2-Nitrophenoxy)acetic acid ≥98.0% (by GC, titration analysis)
(2-Nitrophenoxy)acetic acid

CAS Number: 1878-87-1
MDL Number: MFCD00016993
Molecular Formula: C8H7NO5
Molecular Weight: 197.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 157

Formula: (C₆H₄NO₂)OCH₂COOH
MW: 197.15 g/mol
Melting Pt: 157 °C
MDL Number: MFCD00016993
CAS Number: 1878-87-1
EINECS: 217-527-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b2053a483a9f98e8eb8faffceade7f93ed41630&vid=6b2053a483a9f98e8eb8faffceade7f93ed41630&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR