4-Nitrophenol ≥99.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 1-Hydroxy-4-nitrobenzene, p-Nitrophenol

N0220-500G N0220-25G N0220-100G
TCN0220-500GEA 123.13 USD
TCN0220-500G TCN0220-25G TCN0220-100G
4-Nitrophenol ≥99.0% (by GC)
4-Nitrophenol

CAS Number: 100-02-7
MDL Number: MFCD00007331
Molecular Formula: C6H5NO3
Molecular Weight: 139.11
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 279
Melting point (°C): 114
Flash Point (°C): 169

Formula: C₆H₅NO₃
Boiling Pt: 279 °C
Melting Pt: 114 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD00007331
CAS Number: 100-02-7
EINECS: 202-811-7
UN: 1663
ADR: 6.1,III
Merck Index: 13,06654
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fd741f985998363d3f13c7c26d9dc6e91a5deb9&vid=65d1b5dadddc409031c10f3167ca4d03447d7591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 112.0 to 116.0 deg-C
Solubility in EtOH almost transparency
Transition interval (pH) (very pale yellow)4.8 to 7.6 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR