4-Nitrobenzoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: p-Nitrobenzoyl chloride

N0176-500G N0176-25G
TCN0176-500GEA 97.65 USD
TCN0176-500G TCN0176-25G
4-Nitrobenzoyl chloride ≥98.0% (by GC, titration analysis)
4-Nitrobenzoyl chloride

CAS Number: 122-04-3
MDL Number: MFCD00007345
Molecular Formula: C7H4ClNO3
Molecular Weight: 185.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 128
Melting point (°C): 73
Flash Point (°C): 102

Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Boiling Pt: 128 °C
Melting Pt: 73 °C
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00007345
CAS Number: 122-04-3
EINECS: 204-517-4
UN: 3261
ADR: 8,II
Merck Index: 12,06687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e336ee685243ead2a12eb2041ec21c6bae13877&vid=5e336ee685243ead2a12eb2041ec21c6bae13877&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR