3-Nitrobenzoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: m-Nitrobenzoyl chloride

N0174-500G N0174-25G
TCN0174-500GEA 244 USD
TCN0174-500G TCN0174-25G
3-Nitrobenzoyl chloride ≥98.0% (by GC, titration analysis)
3-Nitrobenzoyl chloride

CAS Number: 121-90-4
MDL Number: MFCD00007247
Molecular Formula: C7H4ClNO3
Molecular Weight: 185.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 27

Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 30 - 33 °C
Density: 1.42 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007247
CAS Number: 121-90-4
EINECS: 204-505-9
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de501305e3b3547ca656e3a8b344384f42aafd4b&vid=de501305e3b3547ca656e3a8b344384f42aafd4b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR