3-Nitrobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Nitrobenzene-1-sulphonyl chloride

N0143-500G N0143-25G
TCN0143-500GEA 332 USD
TCN0143-500G TCN0143-25G
3-Nitrobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
3-Nitrobenzenesulphonyl chloride

CAS Number: 121-51-7
MDL Number: MFCD00007435
Molecular Formula: C6H4ClNO4S
Molecular Weight: 221.61
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 62
Flash Point (°C): 110

Formula: C₆H₄ClNO₄S
MW: 221.62 g/mol
Melting Pt: 62 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007435
CAS Number: 121-51-7
EINECS: 204-476-2
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=943d7daab66e138d76b6f0e812d2ec512c8b2aed&vid=943d7daab66e138d76b6f0e812d2ec512c8b2aed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR