2-Nitrobenzenesulfonyl chloride ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 2-Nitrobenzene-1-sulphonyl chloride

N0142-500G N0142-25G N0142-100G
TCN0142-500GEA 319 USD
TCN0142-500G TCN0142-25G TCN0142-100G
2-Nitrobenzenesulfonyl chloride ≥95.0% (by GC, titration analysis)
2-Nitrobenzenesulphonyl chloride

CAS Number: 1694-92-4
MDL Number: MFCD00007430
Molecular Formula: C6H4ClNO4S
Molecular Weight: 221.61
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 64


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Bioscience, Polyamine Synthesis Reagents
Other Product Names / Synonyms: o-Ns-Cl

Formula: C₆H₄ClNO₄S
MW: 221.62 g/mol
Melting Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00007430
CAS Number: 1694-92-4
EINECS: 216-907-1
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc5fd4c12b577a5b279edd48bd46a968795cef0d&vid=fc5fd4c12b577a5b279edd48bd46a968795cef0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR