4-Nitroanisole ≥98.0%

Supplier: TCI America

N0126-500G N0126-25G N0126-100G
TCN0126-500GEA 124 USD
TCN0126-500G TCN0126-25G TCN0126-100G
4-Nitroanisole ≥98.0%
4-Nitroanisole

CAS Number: 100-17-4
MDL Number: MFCD00007327
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 274
Melting point (°C): 54
Flash Point (°C): 130


Other Product Names / Synonyms: 1-Methoxy-4-nitrobenzene

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 274 °C
Melting Pt: 54 °C
Density: 1.254 g/cm³ (20 °C)
Flash Pt: 130 °C
MDL Number: MFCD00007327
CAS Number: 100-17-4
EINECS: 202-825-3
UN: 3458
ADR: 6.1,III
Merck Index: 12,06682
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b1aa84a00db3bf24653bf33a0c2ac555a94a3a8&vid=443d21b20f206a91b47f6dee9c47ce8bdcc2876a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR