4-Nitroaniline ≥98.0%

Supplier: TCI America
Danger

Synonyms: p-Nitroaniline, pNA

N0119-500G N0119-25G
TCN0119-500GEA 186.14 USD
TCN0119-500G TCN0119-25G
4-Nitroaniline ≥98.0%
4-Nitroaniline

CAS Number: 100-01-6
MDL Number: MFCD00007858
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 332
Melting point (°C): 148
Flash Point (°C): 199

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Boiling Pt: 332 °C
Melting Pt: 148 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00007858
CAS Number: 100-01-6
EINECS: 202-810-1
UN: 1661
ADR: 6.1,II
Merck Index: 12,06681
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=004b7dd75e92352cf4b7e738e089a060d9ceea37&vid=477c79e71f2ffbb78f5cf0ac8ccb29d032461a18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR