Nicotinaldehyde ≥98.0%
Supplier: TCI America
Synonyms:
Pyridine-3-carboxaldehyde, 3-Pyridinecarboxaldehyde, 3-Pyridinecarbaldehyde
N0090-25ML
N0090-100ML
TCN0090-500MLEA
636.54
USD
TCN0090-500ML
TCN0090-25ML
TCN0090-100ML
Nicotinaldehyde ≥98.0%
Nicotinaldehyde
CAS Number: 500-22-1
MDL Number: MFCD00006382
Molecular Formula: C6H5NO
Molecular Weight: 107.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): 43
Specific Gravity (20/20): 1.14
Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction)
Other Product Names / Synonyms: Nicotinaldehyde
Formula:
C₆H₅NO MW: 107.11 g/mol Boiling Pt: 97 °C Density: 1.14 (20 °C) Flash Pt: 43 °C |
MDL Number:
MFCD00006382 CAS Number: 500-22-1 EINECS: 207-900-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...