1-Naphthol ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene
N0026-25G
N0026-100G
TCN0026-25GEA
34.19
USD
TCN0026-25G
TCN0026-100G
1-Naphthol ≥99.0%
1-Naphthol
CAS Number: 90-15-3
MDL Number: MFCD00003930
Molecular Formula: C10H8O
Molecular Weight: 144.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 147
Melting point (°C): 96
Flash Point (°C): 125
Lambda max.: 324 nm (MeOH)
Formula:
C₁₀H₈O MW: 144.17 g/mol Boiling Pt: 147 °C Melting Pt: 96 °C Density: 1.224 g/cm³ (20 °C) Flash Pt: 125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003930 CAS Number: 90-15-3 EINECS: 201-969-4 UN: 2811 ADR: 6.1,III Merck Index: 13,06409 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...