Mesityl oxide ≥85.0% (contains 4-Methyl-4-penten-2-one)

Supplier: TCI America
Danger

Synonyms: 4-Methyl-3-penten-2-one

M1340-500ML M1340-25ML
TCM1340-500MLEA 107 USD
TCM1340-500ML TCM1340-25ML
Mesityl oxide ≥85.0% (contains 4-Methyl-4-penten-2-one)
Mesityl oxide

CAS Number: 141-79-7
MDL Number: MFCD00008900
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Flash Point (°C): 25
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: Isopropylideneacetone, Mesityl Oxide

Formula: C₆H₁₀O
MW: 98.14 g/mol
Density: 0.85 (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008900
CAS Number: 141-79-7
EINECS: 205-502-5
UN: 1229
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4665e19a30af7cdd7c1385f35d5d8a047acdf96&vid=d4665e19a30af7cdd7c1385f35d5d8a047acdf96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR