Methyldiphenylphosphine ≥97.0%

Supplier: TCI America

M1318-1G M1318-5G
TCM1318-1GEA 64 USD
TCM1318-1G TCM1318-005G
Methyldiphenylphosphine ≥97.0%
Methyldiphenylphosphine

CAS Number: 1486-28-8
MDL Number: MFCD00008508
Molecular Formula: C13H13P
Molecular Weight: 200.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 186
Flash Point (°C): 110
Specific Gravity (20/20): 1.06

Formula: C₁₃H₁₃P
Boiling Pt: 186 °C
Melting Pt: 28 °C
Density: 1.06 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008508
CAS Number: 1486-28-8
EINECS: 216-065-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=910994813caf4f70ce838fcfa21dd20dad4c0dbd&vid=910994813caf4f70ce838fcfa21dd20dad4c0dbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR