(±)-2-Methylpiperazine ≥99.0%

Supplier: TCI America

Synonyms: 2-Methylpiperazine

M1179-500G M1179-25G
TCM1179-500GEA 297 USD
TCM1179-500G TCM1179-25G
(±)-2-Methylpiperazine ≥99.0%
(±)-2-Methylpiperazine

CAS Number: 109-07-9
MDL Number: MFCD00005954
Molecular Formula: C5H12N2
Molecular Weight: 100.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 153
Melting point (°C): 64
Flash Point (°C): 65

Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 153 °C
Melting Pt: 64 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00005954
CAS Number: 109-07-9
EINECS: 203-644-2
UN: 2925
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b968a4bf8e57e259e3d739f5bdc242dc971978b8&vid=b968a4bf8e57e259e3d739f5bdc242dc971978b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR