3-Methyl-3-penten-2-one ≥95.0%

Supplier: TCI America

Synonyms: 3-Methylpent-3-en-2-one

M1139-500ML M1139-25ML
TCM1139-500MLEA 356 USD
TCM1139-500ML TCM1139-25ML
3-Methyl-3-penten-2-one ≥95.0%
3-Methyl-3-penten-2-one

CAS Number: 565-62-8
MDL Number: MFCD00151839
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 34
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: 2-Acetyl-2-butene

Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 140 °C
Melting Pt: -70 °C
Density: 0.88 (20 °C)
Flash Pt: 34 °C
MDL Number: MFCD00151839
CAS Number: 565-62-8
EINECS: 209-283-7
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cbcb7c81a46a094be216d0f5d9e8e328a582eea&vid=7cbcb7c81a46a094be216d0f5d9e8e328a582eea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR