3-Methyl-2-cyclopenten-1-one ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Methylcyclopent-2-enone, 3-Methyl-2-cyclopentenone

M0956-25ML M0956-100ML
TCM0956-025MLEA 169 USD
TCM0956-025ML TCM0956-100ML
3-Methyl-2-cyclopenten-1-one ≥98.0% (by GC)
3-Methyl-2-cyclopenten-1-one

CAS Number: 2758-18-1
MDL Number: MFCD00001403
Molecular Formula: C6H8O
Molecular Weight: 96.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 65
Specific Gravity (20/20): 0.98

Formula: C₆H₈O
MW: 96.13 g/mol
Boiling Pt: 108 °C
Melting Pt: 2 °C
Density: 0.98 (20 °C)
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001403
CAS Number: 2758-18-1
EINECS: 220-421-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9731a583f8ed11754e45baea4871396d9a5bf12d&vid=e556c06239b183b432e2dca0f509387e164fe1f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR