5-Methoxy-m-cresol ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
3-Hydroxy-5-methylanisol, 1-Hydroxy-3-methoxy-5-methylbenzene, 3-Methoxy-5-methylphenol, 3-Hydroxy-5-methoxytoluene
M0895-1G
M0895-5G
TCM0895-001GEA
44
USD
TCM0895-001G
TCM0895-005G
5-Methoxy-m-cresol ≥98.0%
5-Methoxy-m-cresol
CAS Number: 3209-13-0
MDL Number: MFCD00059261
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 63
Other Product Names / Synonyms: 5-Methoxy-m-cresol, Orcinol Monomethyl Ether
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Melting Pt: 63 °C |
MDL Number:
MFCD00059261 CAS Number: 3209-13-0 EINECS: 221-716-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...