Methyl-6-quinolyl ether ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 6-Methoxyquinoline

M0891-25G M0891-5G
TCM0891-025GEA 279 USD
TCM0891-025G TCM0891-5G
Methyl-6-quinolyl ether ≥97.0% (by GC, titration analysis)
Methyl-6-quinolyl ether

CAS Number: 5263-87-6
MDL Number: MFCD00006800
Molecular Formula: C10H9NO
Molecular Weight: 159.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 284
Flash Point (°C): 104
Freezing point (°C): 22


Other Product Names / Synonyms: p-Quinanisole

Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 284 °C
Melting Pt: 19 °C
Density: 1.152 g/cm³ (20 °C)
Flash Pt: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006800
CAS Number: 5263-87-6
EINECS: 226-077-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0f2071d9ac9d56cafee43b90b2f545cbbed8137&vid=b0f2071d9ac9d56cafee43b90b2f545cbbed8137&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR