(Methylamino)acetaldehyde dimethylacetal ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M0884-500ML M0884-25ML
TCM0884-500MLEA 513 USD
TCM0884-500ML TCM0884-025ML
(Methylamino)acetaldehyde dimethylacetal ≥98.0% (by GC, titration analysis)
(Methylamino)acetaldehyde dimethylacetal

CAS Number: 122-07-6
MDL Number: MFCD00008485
Molecular Formula: C5H13NO2
Molecular Weight: 119.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 29
Specific Gravity (20/20): 0.93


Other Product Names / Synonyms: N-(2,2-Dimethoxyethyl)methylamine

Formula: C₅H₁₃NO₂
Boiling Pt: 140 °C
Density: 0.93 (20 °C)
Flash Pt: 29 °C
MDL Number: MFCD00008485
CAS Number: 122-07-6
EINECS: 204-520-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74a344a03d7c696362020875a83f6db0b9d60509&vid=74a344a03d7c696362020875a83f6db0b9d60509&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR