(4-Methoxyphenyl)acetonitrile ≥97.0%

Supplier: TCI America

Synonyms: 4-Methoxybenzyl cyanide, 4-Methoxyphenylacetonitrile

M0856-500G M0856-25G
TCM0856-500GEA 265 USD
TCM0856-500G TCM0856-25G
(4-Methoxyphenyl)acetonitrile ≥97.0%
(4-Methoxyphenyl)acetonitrile

CAS Number: 104-47-2
MDL Number: MFCD00001919
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 290
Flash Point (°C): 117
Freezing point (°C): 8
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: 4-Methoxybenzyl Cyanide

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 290 °C
Melting Pt: 8 °C
Density: 1.09 (20 °C)
Flash Pt: 117 °C
MDL Number: MFCD00001919
CAS Number: 104-47-2
EINECS: 203-206-0
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db7859754b948a613db6fc70e43deff8bd1a28ec&vid=db7859754b948a613db6fc70e43deff8bd1a28ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR