4-Methoxybenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Methoxybenzene-1-sulphonyl chloride

M0802-25G M0802-100G
TCM0802-25GEA 60 USD
TCM0802-25G TCM0802-100G
4-Methoxybenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
4-Methoxybenzenesulphonyl chloride

CAS Number: 98-68-0
MDL Number: MFCD00007446
Molecular Formula: C7H7ClO3S
Molecular Weight: 206.64
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 173
Flash Point (°C): 110
Freezing point (°C): 39


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: p-Anisolesulfonyl Chloride, 4-Methoxyphenylsulfonyl Chloride

Formula: C₇H₇ClO₃S
MW: 206.65 g/mol
Boiling Pt: 173 °C
Melting Pt: 40…42 °C
Flash Pt: 110 °C
MDL Number: MFCD00007446
CAS Number: 98-68-0
EINECS: 202-692-1
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a32eb201e6856f65b34fa00b45769f4504b2940&vid=1a32eb201e6856f65b34fa00b45769f4504b2940&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR