Butyltin hydroxide oxide
Supplier: TCI America
M0744-25G
M0744-500G
TCM0744-025GEA
41
USD
TCM0744-025G
TCM0744-500G
Butyltin hydroxide oxide
Butyltin hydroxide oxide
CAS Number: 2273-43-0
MDL Number: MFCD00013927
Molecular Formula: C4H10O2Sn
Molecular Weight: 208.83
Form: Crystal
Color: White
Flash Point (°C): 80
Other Product Names / Synonyms: Butylenestannonic Acid
Formula:
C₄H₁₀O₂Sn Flash Pt: 80 °C |
MDL Number:
MFCD00013927 CAS Number: 2273-43-0 EINECS: 218-880-1 UN: 3146 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...