(4-Methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 4-Methoxyphenylacetic acid

M0742-25G M0742-500G
TCM0742-025GEA 20.35 USD
TCM0742-025G TCM0742-500G
(4-Methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
(4-Methoxyphenyl)acetic acid

CAS Number: 104-01-8
MDL Number: MFCD00004345
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 305
Melting point (°C): 86


Other Product Names / Synonyms: Homoanisic Acid

Formula: (C₆H₄(OCH₃))CH₂COOH
MW: 166.18 g/mol
Boiling Pt: 305 °C
Melting Pt: 86 °C
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00004345
CAS Number: 104-01-8
EINECS: 203-166-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60fbbf34c08a9201aa19971b12f2636ceaf79b13&vid=60fbbf34c08a9201aa19971b12f2636ceaf79b13&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR