2,2,2-Trifluoroethyl methacrylate ≥98.0% stabilised

Supplier: TCI America

Synonyms: Methacrylic acid 2,2,2-trifluoroethyl ester

M0738-25ML M0738-500ML M0738-100ML
TCM0738-025MLEA 30.3 USD
TCM0738-025ML TCM0738-500ML TCM0738-100ML
2,2,2-Trifluoroethyl methacrylate ≥98.0% stabilised
2,2,2-Trifluoroethyl methacrylate

(stabilized with MEHQ)
CAS Number: 352-87-4
MDL Number: MFCD00013576
Molecular Formula: C6H7F3O2
Molecular Weight: 168.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 25
Specific Gravity (20/20): 1.18


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid 2,2,2-Trifluoroethyl Ester

Formula: C₆H₇F₃O₂
MW: 168.12 g/mol
Boiling Pt: 102 °C
Density: 1.18 (20 °C)
Flash Pt: 25 °C
MDL Number: MFCD00013576
CAS Number: 352-87-4
EINECS: 206-525-3
UN: 3272
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f701dbecb6eaccaad953e95fb28be79399ac8f99&vid=f701dbecb6eaccaad953e95fb28be79399ac8f99&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR