3-(Methylthio)-1-propanol ≥99.0%
Supplier: TCI America
M0735-25G
M0735-5G
TCM0735-010GEA
34.41
USD
TCM0735-010G
TCM0735-025G
TCM0735-5G
3-(Methylthio)-1-propanol ≥99.0%
3-(Methylthio)-1-propanol
CAS Number: 505-10-2
MDL Number: MFCD00036560
Molecular Formula: C4H10OS
Molecular Weight: 106.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 87
Specific Gravity (20/20): 1.03
Other Product Names / Synonyms: 3-Hydroxypropyl Methyl Sulfide, 3-Methylmercaptopropyl Alcohol, 3-Methylthio-1-propanol
Formula:
C₄H₁₀OS Boiling Pt: 136 °C Density: 1.03 (20 °C) Flash Pt: 87 °C |
MDL Number:
MFCD00036560 CAS Number: 505-10-2 EINECS: 208-004-6 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...