4-Anisoyl chloride ≥99.0%

Supplier: TCI America
Danger

Synonyms: 4-Methoxybenzoyl chloride

M0721-500G M0721-25G M0721-100G
TCM0721-500GEA 356 USD
TCM0721-500G TCM0721-25G TCM0721-100G
4-Anisoyl chloride ≥99.0%
4-Anisoyl chloride

CAS Number: 100-07-2
MDL Number: MFCD00000687
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Flash Point (°C): 87
Freezing point (°C): 23


Product Categories: Synthetic Organic Chemistry, Nucleic Acid Synthesis, Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method), UV Detection (HPLC Labeling Reagents), UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: p-Anisoyl Chloride

Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 263
Melting Pt: 82…83 °C
Density: 1.261 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000687
CAS Number: 100-07-2
EINECS: 202-816-4
UN: 1729
ADR: 8 II
Merck Index: 13,00676
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d94c46829243640e97a4e1c5375079e5952d546f&vid=d94c46829243640e97a4e1c5375079e5952d546f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR