Maltol ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-Hydroxy-2-methyl-γ-pyrone, Maltol, 3-Hydroxy-2-methyl-4-pyranone, 3-Hydroxy-2-methyl-4H-pyran-4-one, 3-Hydroxy-2-methyl-4-pyrone

M0673-500G M0673-25G M0673-100G
TCM0673-500GEA 272 USD
TCM0673-500G TCM0673-025G TCM0673-100G
Maltol ≥99.0% (by HPLC, titration analysis)
Maltol

CAS Number: 118-71-8
MDL Number: MFCD00006578
Molecular Formula: C6H6O3
Molecular Weight: 126.11
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 162


Other Product Names / Synonyms: Maltol

Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 162 °C
Density: 1.453 g/cm³ (20 °C)
MDL Number: MFCD00006578
CAS Number: 118-71-8
EINECS: 204-271-8
Merck Index: 13,05735
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ae22f9fa0843021eb210c59ea88b6e15c4279c0&vid=3ae22f9fa0843021eb210c59ea88b6e15c4279c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR