1,3-Benzodioxole ≥99.0%

Supplier: TCI America
Warning

Synonyms: Benzo[d][1,3]dioxole, 1,2-Methylenedioxybenzene

M0658-500ML M0658-25ML M0658-100ML
TCM0658-500MLEA 199 USD
TCM0658-500ML TCM0658-25ML TCM0658-100ML
1,3-Benzodioxole ≥99.0%
1,3-Benzodioxole

CAS Number: 274-09-9
MDL Number: MFCD00005818
Molecular Formula: C7H6O2
Molecular Weight: 122.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 172
Melting point (°C): -10
Flash Point (°C): 60
Specific Gravity (20/20): 1.19


Other Product Names / Synonyms: 1,3-Benzodioxole

Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 172 °C
Melting Pt: –10 °C
Density: 1.19 (20 °C)
Flash Pt: 60 °C
MDL Number: MFCD00005818
CAS Number: 274-09-9
EINECS: 205-992-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbfbe0c04c76d5ee1da39db5abeb9fa88ce508f6&vid=bbfbe0c04c76d5ee1da39db5abeb9fa88ce508f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR