3-Methoxypropiononitrile ≥99.0%

Supplier: TCI America
Warning

Synonyms: 3-Methoxypropanenitrile, 3-Methoxypropionitrile

M0653-25ML M0653-500ML
TCM0653-025MLEA 26 USD
TCM0653-025ML TCM0653-500ML
3-Methoxypropiononitrile ≥99.0%
3-Methoxypropiononitrile

CAS Number: 110-67-8
MDL Number: MFCD00001958
Molecular Formula: C4H7NO
Molecular Weight: 85.11
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 166
Melting point (°C): -63
Flash Point (°C): 65
Specific Gravity (20/20): 0.94

Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 166 °C
Melting Pt: –63 °C
Density: 0.94 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00001958
CAS Number: 110-67-8
EINECS: 203-790-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad440cb5d541fa76a65600f9bb12c9511fd0b2a0&vid=ad440cb5d541fa76a65600f9bb12c9511fd0b2a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR