4-Methylimidazole ≥98.0%

Supplier: TCI America

Synonyms: 4-Methyl-1H-imidazole

M0636-25G M0636-100G
TCM0636-25GEA 38.67 USD
TCM0636-25G TCM0636-100G
4-Methylimidazole ≥98.0%
4-Methylimidazole

CAS Number: 822-36-6
MDL Number: MFCD00005201
Molecular Formula: C4H6N2
Molecular Weight: 82.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 263
Melting point (°C): 52
Flash Point (°C): 157

Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 263 °C
Melting Pt: 52 °C
Density: 1.0416 g/cm³ (14.3 °C)
Flash Pt: 157 °C
MDL Number: MFCD00005201
CAS Number: 822-36-6
EINECS: 212-497-3
UN: 3263
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4329e38c72e5243831bf2d392af9be3f92504d07&vid=4329e38c72e5243831bf2d392af9be3f92504d07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR