2-Methylthiophene ≥97.0%

Supplier: TCI America
Danger

M0635-500ML M0635-25ML
TCM0635-500MLEA 962 USD
TCM0635-500ML TCM0635-25ML
2-Methylthiophene ≥97.0%
2-Methylthiophene

CAS Number: 554-14-3
MDL Number: MFCD00005451
Molecular Formula: C5H6S
Molecular Weight: 98.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Melting point (°C): -63
Flash Point (°C): 8
Specific Gravity (20/20): 1.02

Formula: C₅H₆S
Boiling Pt: 113 °C
Melting Pt: -63 °C
Density: 1.02 (20 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00005451
CAS Number: 554-14-3
EINECS: 209-063-0
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5924a4e99bd0ff08c135f2e8b0ad3ca86367edcf&vid=5924a4e99bd0ff08c135f2e8b0ad3ca86367edcf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR