(±)-2-Methylvaleraldehyde ≥95.0%
Supplier: TCI America
Synonyms:
2-Methylpentanal, 2-Methylvaleraldehyde
M0596-500ML
M0596-25ML
TCM0596-500MLEA
110
USD
TCM0596-500ML
TCM0596-25ML
(±)-2-Methylvaleraldehyde ≥95.0%
(±)-2-Methylvaleraldehyde
CAS Number: 123-15-9
MDL Number: MFCD00006986
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 119
Flash Point (°C): 20
Specific Gravity (20/20): 0.81
Other Product Names / Synonyms: 2-Methylpentanal
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 119 °C Melting Pt: -100 °C Density: 0.81 (20 °C) Flash Pt: 20 °C |
MDL Number:
MFCD00006986 CAS Number: 123-15-9 EINECS: 204-605-2 UN: 2367 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...