(±)-2,3-Epoxypropyl methacrylate ≥95.0% stabilized
Supplier: TCI America
|
Warning
|
Synonyms:
Oxiran-2-ylmethyl methacrylate, Glycidyl methacrylate
Stabilization:
Stabilised with Stabilised
(stabilized with MEHQ)
CAS Number: 106-91-2
MDL Number: MFCD00005137
Molecular Formula: C7H10O3
Molecular Weight: 142.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 84
Freezing point (°C): -42
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Monomers, Monomers
Other Product Names / Synonyms: Methacrylic Acid Glycidyl Ester
Formula:
C₇H₁₀O₃ MW: 142.15 g/mol Boiling Pt: 189 °C Density: 1.08 (20 °C) Flash Pt: 84 °C |
MDL Number:
MFCD00005137 CAS Number: 106-91-2 EINECS: 203-441-9 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...