3-Methyl-1H-pyrazole ≥97.0%

Supplier: TCI America

M0564-25G M0564-5G
TCM0564-025GEA 71.1 USD
TCM0564-025G TCM0564-005G
3-Methyl-1H-pyrazole ≥97.0%
3-Methyl-1H-pyrazole

CAS Number: 1453-58-3
MDL Number: MFCD00005240
Molecular Formula: C4H6N2
Molecular Weight: 82.11
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 204
Flash Point (°C): 93
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: 3-Methyl-1,2-diazole

Formula: C₄H₆N₂
Boiling Pt: 204 °C
Density: 1.02 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00005240
CAS Number: 1453-58-3
EINECS: 215-925-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90849cb5d35eb7e4c901bd7982828690430c9a63&vid=90849cb5d35eb7e4c901bd7982828690430c9a63&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR