1-Methylpiperazine ≥98.0%

Supplier: TCI America

M0553-500ML M0553-25ML M0553-100ML
TCM0553-500MLEA 131 USD
TCM0553-500ML TCM0553-25ML TCM0553-100ML
1-Methylpiperazine ≥98.0%
1-Methylpiperazine

CAS Number: 109-01-3
MDL Number: MFCD00005966
Molecular Formula: C5H12N2
Molecular Weight: 100.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Melting point (°C): -5
Flash Point (°C): 42
Specific Gravity (20/20): 0.91

Formula: C₅H₁₂N₂
Boiling Pt: 137 °C
Melting Pt: –5 °C
Density: 0.91 (20 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00005966
CAS Number: 109-01-3
EINECS: 203-639-5
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a6a8acf40824db0ae74d15c5399ab06910c7653&vid=3a6a8acf40824db0ae74d15c5399ab06910c7653&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR