L(-)-Menthol ≥99.0%

Supplier: TCI America
Warning

Synonyms: (1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, L-Menthol, Levomenthol, L(-)-Menthyl alcoholol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R)-(-)-Menthol, (-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1R,2S,5R)-(-)-Menthol

M0545-25G M0545-500G
TCM0545-025GEA 28.96 USD
TCM0545-025G TCM0545-500G
L(-)-Menthol ≥99.0%
L(-)-Menthol

CAS Number: 2216-51-5
MDL Number: MFCD00062979
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 216
Melting point (°C): 43
Flash Point (°C): 85
Specific rotation [a]20/D: -50 deg (C=10, EtOH)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Monocyclic Monoterpenes
Other Product Names / Synonyms: (-)-p-Menthan-3-ol

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 212
Melting Pt: 43 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00062979
CAS Number: 2216-51-5
EINECS: 218-690-9
Merck Index: 13,05861
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d56f87b67627ce9cdfb1a9a8b9e0b763008cccc&vid=7d56f87b67627ce9cdfb1a9a8b9e0b763008cccc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR