1,1,3,3-Tetramethoxypropane ≥98.0%

Supplier: TCI America

Synonyms: Malonaldehyde bis(dimethyl acetal)

M0544-25G M0544-500G
TCM0544-025GEA 17 USD
TCM0544-025G TCM0544-500G
1,1,3,3-Tetramethoxypropane ≥98.0%
1,1,3,3-Tetramethoxypropane

CAS Number: 102-52-3
MDL Number: MFCD00008488
Molecular Formula: C7H16O4
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 63
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Malonaldehyde Bis(dimethyl Acetal)

Formula: C₇H₁₆O₄
MW: 164.2 g/mol
Boiling Pt: 78 °C
Density: 1.00 (20 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00008488
CAS Number: 102-52-3
EINECS: 203-037-2
UN: 3271
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15ab1290cfce2650e852c9e8db42c4fd55a17748&vid=15ab1290cfce2650e852c9e8db42c4fd55a17748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR