3-Methylcrotonic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3,3-Dimethylacrylic acid, 3-Methylbut-2-enoic acid
M0543-500G
M0543-25G
TCM0543-500GEA
154
USD
TCM0543-500G
TCM0543-25G
3-Methylcrotonic acid ≥98.0% (by GC, titration analysis)
3-Methylcrotonic acid
CAS Number: 541-47-9
MDL Number: MFCD00004366
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 98
Melting point (°C): 68
Flash Point (°C): 93
Product Categories: Bioscience, Hemiterpenes
Other Product Names / Synonyms: 3,3-Dimethylacrylic Acid, 3-Methyl-2-butenoic Acid, Senecioic Acid
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 98 °C Melting Pt: 68 °C Flash Pt: 93 °C |
MDL Number:
MFCD00004366 CAS Number: 541-47-9 EINECS: 208-782-7 UN: 3261 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...