1-Methyl-4-piperidone ≥98.0%

Supplier: TCI America

Synonyms: N-Methyl-4-piperidone

M0530-500G M0530-25G M0530-100G
TCM0530-500GEA 258 USD
TCM0530-500G TCM0530-25G TCM0530-100G
1-Methyl-4-piperidone ≥98.0%
1-Methyl-4-piperidone

CAS Number: 1445-73-4
MDL Number: MFCD00006191
Molecular Formula: C6H11NO
Molecular Weight: 113.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 72
Flash Point (°C): 58
Specific Gravity (20/20): 0.97

Formula: C₆H₁₁NO
Boiling Pt: 72 °C
Density: 0.97 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00006191
CAS Number: 1445-73-4
EINECS: 215-895-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc9c7f01590f7cab3042351c1232cacd97f2748e&vid=fc9c7f01590f7cab3042351c1232cacd97f2748e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR