Isopropyl myristate ≥98.0%

Supplier: TCI America

M0481-500ML M0481-25ML
TCM0481-500MLEA 96.78 USD
TCM0481-500ML TCM0481-025ML
Isopropyl myristate ≥98.0%
Isopropyl myristate

CAS Number: 110-27-0
MDL Number: MFCD00008982
Molecular Formula: C17H34O2
Molecular Weight: 270.46
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 193
Flash Point (°C): 164
Freezing point (°C): 7
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: Isopropyl Tetradecanoate, Myristic Acid Isopropyl Ester

Formula: C₁₇H₃₄O₂
Boiling Pt: 193 °C
Melting Pt: 0…1 °C
Density: 0.85 (20 °C)
Flash Pt: 164 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008982
CAS Number: 110-27-0
EINECS: 203-751-4
Merck Index: 13,05235
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29108f1211375af87308e5fe997ebb95385a880e&vid=29108f1211375af87308e5fe997ebb95385a880e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR