Dimethyl sulfide ≥99.0%

Supplier: TCI America
Danger

Synonyms: DMS

M0431-25ML M0431-100ML
TCM0431-25MLEA 21.37 USD
TCM0431-25ML TCM0431-100ML
Dimethyl sulfide ≥99.0%
Dimethyl sulphide

CAS Number: 75-18-3
MDL Number: MFCD00008562
Molecular Formula: C2H6S
Molecular Weight: 62.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 37
Melting point (°C): -98
Flash Point (°C): -34
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: Dimethyl Sulfide, DMS

Formula: C₂H₆S
MW: 62.14 g/mol
Boiling Pt: 37 °C
Melting Pt: -98 °C
Density: 0.85 (20 °C)
Flash Pt: -34 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008562
CAS Number: 75-18-3
EINECS: 200-846-2
UN: 1164
ADR: 3,II
Merck Index: 12,06204
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7feb1883a32e4c25ab1d615c02b5d8d12de2715&vid=e7feb1883a32e4c25ab1d615c02b5d8d12de2715&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR