4-Methylpyrocatechol ≥98.0%
Supplier: TCI America
Synonyms:
4-Methylbenzene-1,2-diol, 4-Methylcatechol
M0413-25G
M0413-500G
TCM0413-025GEA
60
USD
TCM0413-025G
TCM0413-500G
4-Methylpyrocatechol ≥98.0%
4-Methylpyrocatechol
CAS Number: 452-86-8
MDL Number: MFCD00002205
Molecular Formula: C7H8O2
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 117
Melting point (°C): 66
Flash Point (°C): 140
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 1,2-Dihydroxy-4-methylbenzene, 3,4-Dihydroxytoluene, Homocatechol, 4-Methylpyrocatechol
Formula:
C₇H₈O₂ MW: 124.14 g/mol Boiling Pt: 117 °C Melting Pt: 66 °C Flash Pt: 140 °C |
MDL Number:
MFCD00002205 CAS Number: 452-86-8 EINECS: 207-214-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...