Thymol ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
4-Isopropyl-3-hydroxytoluene, 2-Isopropyl-5-methylphenol, 6-Isopropyl-m-cresol
M0410-500G
M0410-25G
TCM0410-500GEA
99.85
USD
TCM0410-500G
TCM0410-25G
Thymol ≥99.0%
Thymol
CAS Number: 89-83-8
MDL Number: MFCD00002309
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 232
Melting point (°C): 51
Flash Point (°C): 115
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Monocyclic Monoterpenes
Other Product Names / Synonyms: 6-Isopropyl-m-cresol, 6-Isopropyl-3-methylphenol
Formula:
2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH MW: 150.22 g/mol Boiling Pt: 232 °C Melting Pt: 51 °C Density: 0.965 g/cm³ (25 °C) Flash Pt: 115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002309 CAS Number: 89-83-8 EINECS: 201-944-8 UN: 2430 ADR: 8,III Merck Index: 13,09476 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...