(±)-4-Methyl-2-pentanol ≥98.0%

Supplier: TCI America
Danger

Synonyms: (±)-Methyl isobutyl carbinol

M0386-500ML M0386-25ML
TCM0386-500MLEA 27.47 USD
TCM0386-500ML TCM0386-25ML
(±)-4-Methyl-2-pentanol ≥98.0%
(±)-4-Methyl-2-pentanol

CAS Number: 108-11-2
MDL Number: MFCD00004550
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Melting point (°C): -90
Flash Point (°C): 43
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: sec-Hexyl Alcohol, Isobutylmethylcarbinol

Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 132 °C
Melting Pt: -90 °C
Density: 0.81 (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00004550
CAS Number: 108-11-2
EINECS: 203-551-7
UN: 2053
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e7725cef1d9497ed2cdd7b577628569163da73c&vid=0e7725cef1d9497ed2cdd7b577628569163da73c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR