2-Methylpentane ≥98.0%

Supplier: TCI America
Danger

M0382-500ML M0382-25ML
TCM0382-500MLEA 175.65 USD
TCM0382-500ML TCM0382-25ML
2-Methylpentane ≥98.0%
2-Methylpentane

CAS Number: 107-83-5
MDL Number: MFCD00009406
Molecular Formula: C6H14
Molecular Weight: 86.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): -20
Specific Gravity (20/20): 0.65


Other Product Names / Synonyms: Isohexane

Formula: C₆H₁₄
Boiling Pt: 60 °C
Melting Pt: –154 °C
Density: 0.65 (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00009406
CAS Number: 107-83-5
EINECS: 203-523-4
UN: 1208
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=469cf67251f313da1eb016500b32132edfc419c4&vid=a651f809c9bc4bedbd4879f52ebb36cba2eef8ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR