5-Methylbenzotriazole ≥99.0% (by titrimetric analysis)

Supplier: TCI America

M0249-500G M0249-25G
TCM0249-500GEA 474 USD
TCM0249-500G TCM0249-025G
5-Methylbenzotriazole ≥99.0% (by titrimetric analysis)
5-Methylbenzotriazole

CAS Number: 136-85-6
MDL Number: MFCD00005702
Molecular Formula: C7H7N3
Molecular Weight: 133.15
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Boiling point (°C): 323
Melting point (°C): 83
Lambda max.: 276 nm (H2O)Storage Temperature: 0-10°C

Formula: C₇H₇N₃
Boiling Pt: 323 °C
Melting Pt: 83 °C
MDL Number: MFCD00005702
CAS Number: 136-85-6
EINECS: 205-265-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=884515db29fda1c7caf325738b97189256c0a020&vid=884515db29fda1c7caf325738b97189256c0a020&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR