4,4'-Methylenedianiline ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: 4,4’-Diaminodiphenylmethane, Bis(4-aminophenyl)methane, 4,4'-Diaminodiphenylmethane, 4,4'-Methylenedianiline

M0220-500G M0220-25G
TCM0220-500GEA 45 USD
TCM0220-500G TCM0220-25G
4,4'-Methylenedianiline ≥98.0% (by titrimetric analysis)
4,4'-Methylenedianiline

CAS Number: 101-77-9
MDL Number: MFCD00007914
Molecular Formula: C13H14N2
Molecular Weight: 198.27
Purity/Analysis Method: >98.0% (T)
Form: Pellet
Boiling point (°C): 257
Melting point (°C): 91
Flash Point (°C): 220


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Methylenedianiline

Formula: C₁₃H₁₄N₂
MW: 198.27 g/mol
Boiling Pt: 257 °C
Melting Pt: 91 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00007914
CAS Number: 101-77-9
EINECS: 202-974-4
UN: 2651
ADR: 6.1,III
Merck Index: 13,03000
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ab28932eee832fa367723a9c5fb2609a9512bad&vid=4ab28932eee832fa367723a9c5fb2609a9512bad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR