2-Methyl-1,3-dioxolane ≥98.0%

Supplier: TCI America

M0210-500G M0210-25G
TCM0210-500GEA 616 USD
TCM0210-500G TCM0210-25G
2-Methyl-1,3-dioxolane ≥98.0%
2-Methyl-1,3-dioxolane

CAS Number: 497-26-7
MDL Number: MFCD00003210
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 83
Flash Point (°C): -2
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Acetaldehyde Ethylene Acetal

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 83 °C
Density: 0.98 (20 °C)
Flash Pt: -2 °C
MDL Number: MFCD00003210
CAS Number: 497-26-7
EINECS: 207-841-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eef426aa1262c064a3522a7b61ad95834914fce8&vid=eef426aa1262c064a3522a7b61ad95834914fce8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR