2-Methylcyclohexanone ≥96.0%

Supplier: TCI America
Danger

M0197-25ML M0197-100ML
TCM0197-25MLEA 45.76 USD
TCM0197-25ML TCM0197-100ML
2-Methylcyclohexanone ≥96.0%
2-Methylcyclohexanone

CAS Number: 583-60-8
MDL Number: MFCD00001635
Molecular Formula: C7H12O
Molecular Weight: 112.17
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 48
Specific Gravity (20/20): 0.93

Formula: C₇H₁₂O
Boiling Pt: 165 °C
Melting Pt: –14 °C
Density: 0.93 (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00001635
CAS Number: 583-60-8
EINECS: 209-513-6
UN: 2297
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef6e2cc50168ef11a67b41a83effac2186af1e04&vid=ef6e2cc50168ef11a67b41a83effac2186af1e04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR