Isovaleric acid ≥99.0%

Supplier: TCI America
Danger

Synonyms: 3-Methylbutyric acid, Isopropyl acetic acid, 3-Methylbutanoic acid

M0182-500ML M0182-25ML
TCM0182-500MLEA 52 USD
TCM0182-500ML TCM0182-25ML
Isovaleric acid ≥99.0%
Isovaleric acid

CAS Number: 503-74-2
MDL Number: MFCD00002726
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 79
Specific Gravity (20/20): 0.93


Product Categories: Bioscience, Hemiterpenes
Other Product Names / Synonyms: 3-Methylbutyric Acid

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 175 °C
Melting Pt: -33 °C
Density: 0.93 (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00002726
CAS Number: 503-74-2
EINECS: 207-975-3
UN: 3265
ADR: 8,II
Merck Index: 12,05251
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5332f6fcd6a19ddb47f10972fdf65c46398d1a0&vid=a5332f6fcd6a19ddb47f10972fdf65c46398d1a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR